missing translation for 'onlineSavingsMsg'
Learn More

1-Benzylimidazole 98.0+%, TCI America™

Catalog No. p-7122917
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
B33875G 5 g
B338725G 25 g
2 options

Catalog No. B33875G

Supplier: TCI America B33875G

Only null left

Chemical Identifiers

CAS 4238-71-5
Molecular Formula C10H10N2
Molecular Weight (g/mol) 158.20
MDL Number MFCD00005296
InChI Key KKKDZZRICRFGSD-UHFFFAOYSA-N
Synonym 1-benzyl-1h-imidazole, n-benzylimidazole, imidazole, 1-benzyl, benzylimidazole, 1h-imidazole, 1-phenylmethyl, 1-benzyl imidazole, ccris 5821, chembl14192, 1bn, 1-benzylimdazole
PubChem CID 77918
IUPAC Name 1-benzyl-1H-imidazole
SMILES C(N1C=CN=C1)C1=CC=CC=C1

Specifications

Melting Point 73°C
Boiling Point 122°C
Quantity 5 g
Formula Weight 158.20
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Benzylimidazole