missing translation for 'onlineSavingsMsg'
Learn More

1-Benzylimidazole, 98+%

Catalog No. p-7043442
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0080206 5 g
AAL0080214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0080206 Supplier Thermo Scientific Chemicals Supplier No. L0080206
Only null left

CAS: 4238-71-5 | C10H10N2 | 158.20 g/mol

1-Benzylimidazole acts as an inducer of various cytochrome P-450 isozymes and inhibitor of thromboxane A2 synthase. 1-Benzylimidazole has been used to prepare cyclodextrin-ionic liquid polymer (βCD-BIMOTs-TDI)

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Benzylimidazole acts as an inducer of various cytochrome P-450 isozymes and inhibitor of thromboxane A2 synthase. 1-Benzylimidazole has been used to prepare cyclodextrin-ionic liquid polymer (βCD-BIMOTs-TDI)

Solubility
Soluble in methanol, acetone, DMSO, ethanol, dilute acids (freely soluble), and buffers of pH>6.0 (freely soluble). Insoluble in water.

Notes
Store in a cool, dry place. Keep container closed when not in use. Incompatible with Oxidizing agents.

Chemical Identifiers

CAS 4238-71-5
Molecular Formula C10H10N2
Molecular Weight (g/mol) 158.20
MDL Number MFCD00005296
InChI Key KKKDZZRICRFGSD-UHFFFAOYSA-N
Synonym 1-benzyl-1h-imidazole, n-benzylimidazole, imidazole, 1-benzyl, benzylimidazole, 1h-imidazole, 1-phenylmethyl, 1-benzyl imidazole, ccris 5821, chembl14192, 1bn, 1-benzylimdazole
PubChem CID 77918
IUPAC Name 1-benzylimidazole
SMILES C(N1C=CN=C1)C1=CC=CC=C1

Specifications

Melting Point 68°C to 73°C
Boiling Point 310°C
Quantity 5 g
Beilstein 114571
Solubility Information Soluble in methanol,acetone,DMSO,ethanol,dilute acids (freely soluble),and buffers of pH>6.0 (freely soluble). Insoluble in water.
Formula Weight 158.2
Percent Purity ≥98%
Chemical Name or Material 1-Benzylimidazole
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 224-200-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only