missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Benzyl N-(tert-Butoxycarbonyl)-L-glutamate 98.0+%, TCI America™
$190.13 - $656.14
Chemical Identifiers
| CAS | 30924-93-7 |
|---|---|
| Molecular Formula | C17H23NO6 |
| Molecular Weight (g/mol) | 337.372 |
| MDL Number | MFCD00065568 |
| InChI Key | CVZUKWBYQQYBTF-ZDUSSCGKSA-N |
| Synonym | 1-Benzyl N-Boc-L-glutamate, N-Boc-L-glutamic Acid 1-Benzyl Ester, N-(tert-Butoxycarbonyl)-L-glutamic Acid 1-Benzyl Ester, Boc-Glu-OBzl |
| PubChem CID | 153708 |
| IUPAC Name | (4S)-4-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxo-5-phenylmethoxypentanoic acid |
| SMILES | CC(C)(C)OC(=O)NC(CCC(=O)O)C(=O)OCC1=CC=CC=C1 |
Chemical Identifiers
| 30924-93-7 | |
| 337.372 | |
| CVZUKWBYQQYBTF-ZDUSSCGKSA-N | |
| 153708 | |
| CC(C)(C)OC(=O)NC(CCC(=O)O)C(=O)OCC1=CC=CC=C1 |
| C17H23NO6 | |
| MFCD00065568 | |
| 1-Benzyl N-Boc-L-glutamate, N-Boc-L-glutamic Acid 1-Benzyl Ester, N-(tert-Butoxycarbonyl)-L-glutamic Acid 1-Benzyl Ester, Boc-Glu-OBzl | |
| (4S)-4-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxo-5-phenylmethoxypentanoic acid |
Specifications
| 30924-93-7 | |
| White | |
| MFCD00065568 | |
| 1-Benzyl N-Boc-L-glutamate, N-Boc-L-glutamic Acid 1-Benzyl Ester, N-(tert-Butoxycarbonyl)-L-glutamic Acid 1-Benzyl Ester, Boc-Glu-OBzl | |
| CC(C)(C)OC(=O)NC(CCC(=O)O)C(=O)OCC1=CC=CC=C1 | |
| 337.372 | |
| 337.37 | |
| Crystalline Powder |
| 97°C | |
| C17H23NO6 | |
| 5 g | |
| CVZUKWBYQQYBTF-ZDUSSCGKSA-N | |
| (4S)-4-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxo-5-phenylmethoxypentanoic acid | |
| 153708 | |
| ≥98.0% (T) | |
| 1-Benzyl N-(tert-Butoxycarbonyl)-L-glutamate |
Safety and Handling
TSCA : No