accessibility menu, dialog, popup

UserName

CAS RN 30924-93-7

H3CH3CCH3OONH(S)OHOOO

CAS RN 30924-93-7

IUPAC Name: (4S)-5-(benzyloxy)-4-{[(tert-butoxy)carbonyl]amino}-5-oxopentanoic acid
Synonyms: (4S)-5-(benzyloxy)-4-[(tert-butoxycarbonyl)amino]-5-oxopentanoic acid
Molecular Weight (g/mol): 337.37
Molecular Formula: C17H23NO6
InChi Key: CVZUKWBYQQYBTF-ZDUSSCGKSA-N
SMILES: CC(C)(C)OC(=O)N[C@@H](CCC(O)=O)C(=O)OCC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results