missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyl-4-cyano-4-hydroxypiperidine Hydrochloride 98.0+%, TCI America™

Catalog No. B223725G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B223725G 25 g
B22375G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B223725G Supplier TCI America Supplier No. B223725G
Only null left

Chemical Identifiers

CAS 71617-20-4
Molecular Formula C13H17ClN2O
Molecular Weight (g/mol) 252.742
MDL Number MFCD00012836
InChI Key BFCODNKBFLYVOU-UHFFFAOYSA-N
Synonym 1-benzyl-4-cyano-4-hydroxypiperidine hydrochloride, 1-benzyl-4-hydroxypiperidine-4-carbonitrile hydrochloride, 1-benzyl-4-hydroxypiperidine-4-carbonitrile monohydrochloride, 4-hydroxy-1-benzylpiperidine-4-carbonitrile, chloride, acmc-209ojd, 1-benzyl-4-hydroxyisonipecotonitrile hydrochloride, 1-benzyl-4-hydroxy-4-piperidinecarbonitrile hydrochloride, 4-piperidinecarbonitrile,4-hydroxy-1-phenylmethyl-,hydrochloride 1:1
PubChem CID 2723780
IUPAC Name 1-benzyl-4-hydroxypiperidine-4-carbonitrile;hydrochloride
SMILES C1CN(CCC1(C#N)O)CC2=CC=CC=C2.Cl

Specifications

Color White-Yellow
Quantity 25 g
UN Number 3439
Formula Weight 252.74
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Benzyl-4-cyano-4-hydroxypiperidine Hydrochloride