missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Benzyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole 98.0+%, TCI America™
$126.76 - $507.02
Chemical Identifiers
| CAS | 761446-45-1 |
|---|---|
| Molecular Formula | C16H21BN2O2 |
| Molecular Weight (g/mol) | 284.166 |
| MDL Number | MFCD03789252 |
| InChI Key | ZVPORPUUZXIPEF-UHFFFAOYSA-N |
| Synonym | 1-benzyl-1h-pyrazole-4-boronic acid pinacol ester, 1-phenylmethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-benzyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-benzylpyrazole-4-boronic acid pinacol ester, 1-benzyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1-benzyl-4-1h-pyrazoleboronic acid, pinacol ester, 1-benzyl-1h-pyrazole-4-boronic acid, pinacol ester, 1-benzyl-1h-pyrazol-4-ylboronic acid, pinacol ester, 1-benzyl-4-pyrazoleboronic acid pinacol ester, 1-benzyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole |
| PubChem CID | 2734771 |
| IUPAC Name | 1-benzyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole |
| SMILES | B1(OC(C(O1)(C)C)(C)C)C2=CN(N=C2)CC3=CC=CC=C3 |
Chemical Identifiers
| 761446-45-1 | |
| 284.166 | |
| ZVPORPUUZXIPEF-UHFFFAOYSA-N | |
| 2734771 | |
| B1(OC(C(O1)(C)C)(C)C)C2=CN(N=C2)CC3=CC=CC=C3 |
| C16H21BN2O2 | |
| MFCD03789252 | |
| 1-benzyl-1h-pyrazole-4-boronic acid pinacol ester, 1-phenylmethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-benzyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-benzylpyrazole-4-boronic acid pinacol ester, 1-benzyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1-benzyl-4-1h-pyrazoleboronic acid, pinacol ester, 1-benzyl-1h-pyrazole-4-boronic acid, pinacol ester, 1-benzyl-1h-pyrazol-4-ylboronic acid, pinacol ester, 1-benzyl-4-pyrazoleboronic acid pinacol ester, 1-benzyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole | |
| 1-benzyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole |
Specifications
| 761446-45-1 | |
| White | |
| MFCD03789252 | |
| 1-benzyl-1h-pyrazole-4-boronic acid pinacol ester, 1-phenylmethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-benzyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-benzylpyrazole-4-boronic acid pinacol ester, 1-benzyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1-benzyl-4-1h-pyrazoleboronic acid, pinacol ester, 1-benzyl-1h-pyrazole-4-boronic acid, pinacol ester, 1-benzyl-1h-pyrazol-4-ylboronic acid, pinacol ester, 1-benzyl-4-pyrazoleboronic acid pinacol ester, 1-benzyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole | |
| B1(OC(C(O1)(C)C)(C)C)C2=CN(N=C2)CC3=CC=CC=C3 | |
| 284.166 | |
| 284.17 | |
| Crystalline Powder |
| 89°C | |
| C16H21BN2O2 | |
| 1 g | |
| ZVPORPUUZXIPEF-UHFFFAOYSA-N | |
| 1-benzyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole | |
| 2734771 | |
| ≥98.0% (GC) | |
| 1-Benzyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole |
Safety and Handling
TSCA : No