accessibility menu, dialog, popup

UserName

CH3H3COBOH3CH3CNN

CAS RN 761446-45-1

IUPAC Name: 1-benzyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Synonyms: 1-benzyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
Molecular Weight (g/mol): 284.17
Molecular Formula: C16H21BN2O2
InChi Key: ZVPORPUUZXIPEF-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CN(CC2=CC=CC=C2)N=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results