missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole 98.0+%, TCI America™

Catalog No. B43641G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B43641G 1 g
B43645G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B43641G Supplier TCI America Supplier No. B43641G

Chemical Identifiers

CAS 761446-45-1
Molecular Formula C16H21BN2O2
Molecular Weight (g/mol) 284.166
MDL Number MFCD03789252
InChI Key ZVPORPUUZXIPEF-UHFFFAOYSA-N
Synonym 1-benzyl-1h-pyrazole-4-boronic acid pinacol ester, 1-phenylmethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-benzyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-benzylpyrazole-4-boronic acid pinacol ester, 1-benzyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1-benzyl-4-1h-pyrazoleboronic acid, pinacol ester, 1-benzyl-1h-pyrazole-4-boronic acid, pinacol ester, 1-benzyl-1h-pyrazol-4-ylboronic acid, pinacol ester, 1-benzyl-4-pyrazoleboronic acid pinacol ester, 1-benzyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole
PubChem CID 2734771
IUPAC Name 1-benzyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CN(N=C2)CC3=CC=CC=C3

Specifications

Melting Point 89°C
Color White
Quantity 1 g
Formula Weight 284.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Benzyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole