missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyl-3-pyrrolidinol 96.0+%, TCI America™

Catalog No. p-7119142
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B15975G 5 g
B159725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B15975G Supplier TCI America Supplier No. B15975G
Only null left

Chemical Identifiers

CAS 775-15-5
Molecular Formula C11H15NO
Molecular Weight (g/mol) 177.247
MDL Number MFCD00012132
InChI Key YQMXOIAIYXXXEE-UHFFFAOYSA-N
PubChem CID 69889
IUPAC Name 1-benzylpyrrolidin-3-ol
SMILES C1CN(CC1O)CC2=CC=CC=C2

Specifications

Color Yellow
Boiling Point 143°C
Quantity 5 g
Formula Weight 177.25
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Benzyl-3-pyrrolidinol