missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyl-3-(ethylamino)pyrrolidine 96.0+%, TCI America™

Catalog No. B15605G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B15605G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B15605G Supplier TCI America Supplier No. B15605G

Chemical Identifiers

CAS 115445-21-1
Molecular Formula C13H20N2
Molecular Weight (g/mol) 204.317
MDL Number MFCD00191305
InChI Key ZSIUSRJKSLXIJH-UHFFFAOYSA-N
PubChem CID 10330560
IUPAC Name 1-benzyl-N-ethylpyrrolidin-3-amine
SMILES CCNC1CCN(C1)CC2=CC=CC=C2

Specifications

Color Yellow
Quantity 5 g
UN Number 2735
Formula Weight 204.32
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Benzyl-3-(ethylamino)pyrrolidine