missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyl-3-azetidinol, 95%, Thermo Scientific™

Catalog No. p-7038516
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5749803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5749803 Supplier Thermo Scientific Chemicals Supplier No. H5749803
Only null left

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 54881-13-9
Molecular Formula C10H13NO
Molecular Weight (g/mol) 163.22
MDL Number MFCD00963607
InChI Key JOXQHYFVXZZGQZ-UHFFFAOYSA-N
Synonym 1-benzyl-azetidin-3-ol, 1-benzyl-3-hydroxyazetidine, 1-benzyl-3-azetidinol, 1-benzyl 3-hydroxy-azetidine, n-benzyl-3-hydroxyazetidine, 3-azetidinol, 1-phenylmethyl, 1-benzyl azetidine-3-ol, pubchem21322, 1-benzylazetidine-3-ol, 1-benzyl-azetidine-3-ol
PubChem CID 3801345
IUPAC Name 1-benzylazetidin-3-ol
SMILES OC1CN(CC2=CC=CC=C2)C1

Specifications

Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 163.22
Percent Purity 95%
Chemical Name or Material 1-Benzyl-3-azetidinol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures