missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyl-3-aminopyrrolidine 97.0+%, TCI America™

Catalog No. B13545G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B13545G 5 g
B135425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B13545G Supplier TCI America Supplier No. B13545G
Only null left

Chemical Identifiers

CAS 18471-40-4
Molecular Formula C11H16N2
Molecular Weight (g/mol) 176.263
MDL Number MFCD00059036
InChI Key HBVNLKQGRZPGRP-UHFFFAOYSA-N
Synonym 3-Amino-1-benzylpyrrolidine
PubChem CID 2756613
IUPAC Name 1-benzylpyrrolidin-3-amine
SMILES C1CN(CC1N)CC2=CC=CC=C2

Specifications

Color Yellow
Quantity 5 g
UN Number 2735
Formula Weight 176.26
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-Benzyl-3-aminopyrrolidine