missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyl-1-phenylhydrazine Hydrochloride 98.0+%, TCI America™

Catalog No. p-7118442
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B043425G 25 g
B0434500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B043425G Supplier TCI America Supplier No. B043425G

Chemical Identifiers

CAS 5705-15-7
Molecular Formula C13H15ClN2
Molecular Weight (g/mol) 234.73
MDL Number MFCD00050690
InChI Key JTYLHYOCBGPMNO-UHFFFAOYSA-N
Synonym 1-benzyl-1-phenylhydrazine hydrochloride, n-benzyl-n-phenylhydrazine hydrochloride, usaf ek-1666, hydrazine, 1-phenyl-1-phenylmethyl-, monohydrochloride, alpha-benzyl-alpha-phenylhydrazine hydrochloride, n-benzyl-n-phenylhydrazinium 1+ chloride, hydrazine, 1-benzyl-1-phenyl-, hydrochloride, 1-benzyl-1-phenylhydrazine hydrochloride van, hydrazine, 1-phenyl-1-phenylmethyl-, hydrochloride 1:1, acmc-209lvq
PubChem CID 79785
IUPAC Name hydrogen 1-benzyl-1-phenylhydrazine chloride
SMILES [H+].[Cl-].NN(CC1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 176°C
Color White-Yellow
Quantity 25 g
Formula Weight 234.73
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Benzyl-1-phenylhydrazine Hydrochloride
EINECSNumber (3)-0376
RTECSNumber MU9800000
TSCA Yes