missing translation for 'onlineSavingsMsg'
Learn More

1-Benzothiophene-5-carboxylic acid, 97%, Thermo Scientific™

$347.99 - $347.99

Chemical Identifiers

CAS 2060-64-2
Molecular Formula C9H6O2S
Molecular Weight (g/mol) 178.21
MDL Number MFCD04974042
InChI Key SNBYTKLWZRHESA-UHFFFAOYSA-N
Synonym benzo b thiophene-5-carboxylic acid, buttpark 98\04-76, pubchem20599, 5-carboxybenzo b thiophene, benzothiophene-5-carboxylic acid, benzo b thiophene-5-carboxylicacid, 1-benzo b thiophene-5-carboxylic acid
PubChem CID 2795444
IUPAC Name 1-benzothiophene-5-carboxylic acid
SMILES OC(=O)C1=CC=C2SC=CC2=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC24801DA
SDS View Documents
Thermo Scientific Chemicals
CC24801DA
1 g Amber Glass Bottle
Each for $347.99
Only null left
 
Description

Description

Specifications

Chemical Identifiers

2060-64-2
178.21
SNBYTKLWZRHESA-UHFFFAOYSA-N
2795444
OC(=O)C1=CC=C2SC=CC2=C1
C9H6O2S
MFCD04974042
benzo b thiophene-5-carboxylic acid, buttpark 98\04-76, pubchem20599, 5-carboxybenzo b thiophene, benzothiophene-5-carboxylic acid, benzo b thiophene-5-carboxylicacid, 1-benzo b thiophene-5-carboxylic acid
1-benzothiophene-5-carboxylic acid

Specifications

2060-64-2
Amber Glass Bottle
MFCD04974042
benzo b thiophene-5-carboxylic acid, buttpark 98\04-76, pubchem20599, 5-carboxybenzo b thiophene, benzothiophene-5-carboxylic acid, benzo b thiophene-5-carboxylicacid, 1-benzo b thiophene-5-carboxylic acid
OC(=O)C1=CC=C2SC=CC2=C1
178.21
178.21
1-Benzothiophene-5-carboxylic acid
95%
C9H6O2S
1 g
SNBYTKLWZRHESA-UHFFFAOYSA-N
1-benzothiophene-5-carboxylic acid
2795444
95%
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'