CAS RN 2060-64-2
CAS RN 2060-64-2
IUPAC Name:
1-benzothiophene-5-carboxylic acid
Synonyms:
1-benzothiophene-5-carboxylic acid
Molecular Weight (g/mol):
178.21
Molecular Formula:
C9H6O2S
InChi Key:
SNBYTKLWZRHESA-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CC=C2SC=CC2=C1
1
–
3
of
3
results
3
1-Benzothiophene-5-carboxylic acid, 97%, Thermo Scientific™
CAS: 2060-64-2 Molecular Formula: C9H6O2S Molecular Weight (g/mol): 178.21 MDL Number: MFCD04974042 InChI Key: SNBYTKLWZRHESA-UHFFFAOYSA-N Synonym: benzo b thiophene-5-carboxylic acid,buttpark 98\04-76,pubchem20599,5-carboxybenzo b thiophene,benzothiophene-5-carboxylic acid,benzo b thiophene-5-carboxylicacid,1-benzo b thiophene-5-carboxylic acid PubChem CID: 2795444 IUPAC Name: 1-benzothiophene-5-carboxylic acid SMILES: OC(=O)C1=CC=C2SC=CC2=C1