missing translation for 'onlineSavingsMsg'
Learn More

1-Benzothiophene-5-carboxylic acid, 97%, Thermo Scientific™

Catalog No. CC24801DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC24801DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC24801DA Supplier Thermo Scientific Chemicals Supplier No. CC24801DA

Chemical Identifiers

CAS 2060-64-2
Molecular Formula C9H6O2S
Molecular Weight (g/mol) 178.21
MDL Number MFCD04974042
InChI Key SNBYTKLWZRHESA-UHFFFAOYSA-N
Synonym benzo b thiophene-5-carboxylic acid, buttpark 98\04-76, pubchem20599, 5-carboxybenzo b thiophene, benzothiophene-5-carboxylic acid, benzo b thiophene-5-carboxylicacid, 1-benzo b thiophene-5-carboxylic acid
PubChem CID 2795444
IUPAC Name 1-benzothiophene-5-carboxylic acid
SMILES OC(=O)C1=CC=C2SC=CC2=C1

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 178.21
Percent Purity 95%
Chemical Name or Material 1-Benzothiophene-5-carboxylic acid