missing translation for 'onlineSavingsMsg'
Learn More

1-Benzothiophene-3-carboxylic acid, 97%, Thermo Scientific™

Catalog No. CC12301DE
Change view
Click to view available options
Quantity:
5g
10g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC12301DE 5g Amber Glass Bottle
CC12301EA 10g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC12301DE Supplier Thermo Scientific Chemicals Supplier No. CC12301DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5381-25-9
Molecular Formula C9H6O2S
Molecular Weight (g/mol) 178.205
MDL Number MFCD01846406
InChI Key DRBLTQNCQJXSNU-UHFFFAOYSA-N
Synonym benzo b thiophene-3-carboxylic acid, benzothiophene-3-carboxylic acid, benzothiophene-3-carboxylicacid, pubchem13463, acmc-20a10z, ksc178m8d, 1-benzo b thiophene-3-carboxylic acid, #, thianaphthene-3-carboxylic acid
PubChem CID 601280
IUPAC Name 1-benzothiophene-3-carboxylic acid
SMILES C1=CC=C2C(=C1)C(=CS2)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5g
Formula Weight 178.21
Percent Purity 97%
Chemical Name or Material 1-Benzothiophene-3-carboxylic acid
Hazard Category Warning
Hazard Statement GHS P Statement Causes skin irritation. Causes serious eye irritation.
Precautionary Statement GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously