missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Benzothiophen-3-ylmethanol, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC12309DA
Specifications
| 1-Benzothiophen-3-ylmethanol | |
| 96.5 | |
| 97% | |
| C9H8OS | |
| 1g | |
| UYGMKSKKGSUAHB-UHFFFAOYSA-N | |
| 1-benzothiophen-3-ylmethanol | |
| 2776342 | |
| 97% |
| 5381-24-8 | |
| 100.0 | |
| Amber glass bottle | |
| MFCD02682007 | |
| benzo b thiophen-3-ylmethanol, benzo b thiophene-3-methanol, benzothien-3-ylmethanol, benzo, a thiophene-3-methanol, benzo b thiophen-3-yl methanol, 1-benzothiophen-3-yl methanol, 3-hydroxymethylbenzo b thiophene, benzo b thiophen-3-ylmethan-1-ol, 3-hydroxymethyl benzo b thiophene | |
| OCC1=CSC2=CC=CC=C12 | |
| 164.22 | |
| 164.23 |
Chemical Identifiers
| 5381-24-8 | |
| 164.22 | |
| UYGMKSKKGSUAHB-UHFFFAOYSA-N | |
| 2776342 | |
| OCC1=CSC2=CC=CC=C12 |
| C9H8OS | |
| MFCD02682007 | |
| benzo b thiophen-3-ylmethanol, benzo b thiophene-3-methanol, benzothien-3-ylmethanol, benzo, a thiophene-3-methanol, benzo b thiophen-3-yl methanol, 1-benzothiophen-3-yl methanol, 3-hydroxymethylbenzo b thiophene, benzo b thiophen-3-ylmethan-1-ol, 3-hydroxymethyl benzo b thiophene | |
| 1-benzothiophen-3-ylmethanol |