missing translation for 'onlineSavingsMsg'
Learn More

1-Benzothiophen-3-ylmethanol, 97%, Thermo Scientific™

Chemical Identifiers

CAS 5381-24-8
Molecular Formula C9H8OS
Molecular Weight (g/mol) 164.22
MDL Number MFCD02682007
InChI Key UYGMKSKKGSUAHB-UHFFFAOYSA-N
Synonym benzo b thiophen-3-ylmethanol, benzo b thiophene-3-methanol, benzothien-3-ylmethanol, benzo, a thiophene-3-methanol, benzo b thiophen-3-yl methanol, 1-benzothiophen-3-yl methanol, 3-hydroxymethylbenzo b thiophene, benzo b thiophen-3-ylmethan-1-ol, 3-hydroxymethyl benzo b thiophene
PubChem CID 2776342
IUPAC Name (1-benzothiophen-3-yl)methanol
SMILES OCC1=CSC2=CC=CC=C12
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC12309DA
SDS View Documents
Thermo Scientific Chemicals
CC12309DA
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC12309EA
SDS View Documents
Thermo Scientific Chemicals
CC12309EA
10g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

5381-24-8
164.22
UYGMKSKKGSUAHB-UHFFFAOYSA-N
2776342
OCC1=CSC2=CC=CC=C12
C9H8OS
MFCD02682007
benzo b thiophen-3-ylmethanol, benzo b thiophene-3-methanol, benzothien-3-ylmethanol, benzo, a thiophene-3-methanol, benzo b thiophen-3-yl methanol, 1-benzothiophen-3-yl methanol, 3-hydroxymethylbenzo b thiophene, benzo b thiophen-3-ylmethan-1-ol, 3-hydroxymethyl benzo b thiophene
(1-benzothiophen-3-yl)methanol

Specifications

5381-24-8
100.0
Amber glass bottle
MFCD02682007
benzo b thiophen-3-ylmethanol, benzo b thiophene-3-methanol, benzothien-3-ylmethanol, benzo, a thiophene-3-methanol, benzo b thiophen-3-yl methanol, 1-benzothiophen-3-yl methanol, 3-hydroxymethylbenzo b thiophene, benzo b thiophen-3-ylmethan-1-ol, 3-hydroxymethyl benzo b thiophene
OCC1=CSC2=CC=CC=C12
164.22
164.23
1-Benzothiophen-3-ylmethanol
96.5
97%
C9H8OS
1g
UYGMKSKKGSUAHB-UHFFFAOYSA-N
1-benzothiophen-3-ylmethanol
2776342
97%
Videos
SDS
Documents

Documents