missing translation for 'onlineSavingsMsg'
Learn More

1-Benzhydrylpiperazine, 97%

Catalog No. p-7045663
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1153014 25 g
AAL1153022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1153014 Supplier Thermo Scientific Chemicals Supplier No. L1153014
Only null left

CAS: 841-77-0 | C17H20N2 | 252.361 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 841-77-0
Molecular Formula C17H20N2
Molecular Weight (g/mol) 252.361
MDL Number MFCD00038379
InChI Key NWVNXDKZIQLBNM-UHFFFAOYSA-N
Synonym 1-diphenylmethyl piperazine, benzhydrylpiperazine, n-benzhydrylpiperazine, 1-benzhydryl-piperazine, diphenylmethylpiperazine, norcyclizine, normethylcyclizine, piperazine, 1-diphenylmethyl, 4-benzhydrylpiperazine, n-diphenylmethyl piperazine
PubChem CID 70048
IUPAC Name 1-benzhydrylpiperazine
SMILES C1CN(CCN1)C(C2=CC=CC=C2)C3=CC=CC=C3

Specifications

Melting Point 91°C to 92°C
Quantity 25 g
Beilstein 222773
Sensitivity Air Sensitive
Formula Weight 252.36
Percent Purity 97%
Chemical Name or Material 1-Benzhydrylpiperazine
Hazard Category H302
Hazard Statement GHS H Statement
H302-H332-H315-H319-H335
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 212-667-7
RTECSNumber TL6465000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only