missing translation for 'onlineSavingsMsg'
Learn More

1-Benzhydryl-3-azetidinone 96.0+%, TCI America™

Catalog No. B47991G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B47991G 1 g
B47995G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B47991G Supplier TCI America Supplier No. B47991G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 40320-60-3
Molecular Formula C16H15NO
Molecular Weight (g/mol) 237.302
MDL Number MFCD00964539
InChI Key AVUDXLOVIBJFQA-UHFFFAOYSA-N
Synonym 1-diphenylmethyl azetidin-3-one, 1-benzhydryl-azetidin-3-one, 1-1,1-diphenylmethyl azetidin-3-one, 1-diphenylmethyl-3-azetidinone, 1-benzhydryl-3-azetidinone, 3-azetidinone, 1-diphenylmethyl, pubchem10151, acmc-209jd2, n-benzhydrylazetidin-3-one
PubChem CID 3735558
IUPAC Name 1-benzhydrylazetidin-3-one
SMILES C1C(=O)CN1C(C2=CC=CC=C2)C3=CC=CC=C3

Specifications

Melting Point 77°C
Color White-Yellow
Boiling Point 355°C
Quantity 1 g
Formula Weight 237.30
Percent Purity ≥96.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Benzhydryl-3-azetidinone