missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Aza-15-crown 5-Ether 98.0+%, TCI America™
$257.24 - $868.65
Chemical Identifiers
| CAS | 66943-05-3 |
|---|---|
| Molecular Formula | C10H21NO4 |
| Molecular Weight (g/mol) | 219.281 |
| MDL Number | MFCD00075465 |
| InChI Key | BJUOQSZSDIHZNP-UHFFFAOYSA-N |
| Synonym | aza-15-crown-5, 1-aza-15-crown 5-ether, 1-aza-15-crown-5, 1-aza-15-crown, 1-aza-4,7,10,13-tetraoxacyclopentadecane |
| PubChem CID | 544820 |
| IUPAC Name | 1,4,7,10-tetraoxa-13-azacyclopentadecane |
| SMILES | C1COCCOCCOCCOCCN1 |
Chemical Identifiers
| 66943-05-3 | |
| 219.281 | |
| BJUOQSZSDIHZNP-UHFFFAOYSA-N | |
| 544820 | |
| C1COCCOCCOCCOCCN1 |
| C10H21NO4 | |
| MFCD00075465 | |
| aza-15-crown-5, 1-aza-15-crown 5-ether, 1-aza-15-crown-5, 1-aza-15-crown, 1-aza-4,7,10,13-tetraoxacyclopentadecane | |
| 1,4,7,10-tetraoxa-13-azacyclopentadecane |
Specifications
| 66943-05-3 | |
| White | |
| MFCD00075465 | |
| aza-15-crown-5, 1-aza-15-crown 5-ether, 1-aza-15-crown-5, 1-aza-15-crown, 1-aza-4,7,10,13-tetraoxacyclopentadecane | |
| C1COCCOCCOCCOCCN1 | |
| 219.281 | |
| 219.28 | |
| Crystalline Powder |
| 46°C | |
| C10H21NO4 | |
| 1 g | |
| BJUOQSZSDIHZNP-UHFFFAOYSA-N | |
| 1,4,7,10-tetraoxa-13-azacyclopentadecane | |
| 544820 | |
| ≥98.0% (T) | |
| 1-Aza-15-crown 5-Ether |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator