missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Aza-12-crown 4-Ether 98.0+%, TCI America™
$171.34 - $569.85
Chemical Identifiers
| CAS | 41775-76-2 |
|---|---|
| Molecular Formula | C8H18NO3 |
| Molecular Weight (g/mol) | 176.24 |
| MDL Number | MFCD00134269 |
| InChI Key | QNSRHBOZQLXYNV-UHFFFAOYSA-O |
| PubChem CID | 545817 |
| IUPAC Name | 1,4,7-trioxa-10-azacyclododecan-10-ium |
| SMILES | C1COCCOCCOCC[NH2+]1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
A1552250MG
|
TCI America
A1552250MG |
250 mg |
Each for $171.34
|
|
|||||
|
A15521G
|
TCI America
A15521G |
1 g |
Each for $569.85
|
|
|||||
Chemical Identifiers
| 41775-76-2 | |
| 176.24 | |
| QNSRHBOZQLXYNV-UHFFFAOYSA-O | |
| 1,4,7-trioxa-10-azacyclododecan-10-ium |
| C8H18NO3 | |
| MFCD00134269 | |
| 545817 | |
| C1COCCOCCOCC[NH2+]1 |
Specifications
| 41775-76-2 | |
| C8H18NO3 | |
| 250 mg | |
| QNSRHBOZQLXYNV-UHFFFAOYSA-O | |
| 1,4,7-trioxa-10-azacyclododecan-10-ium | |
| 545817 | |
| ≥98.0% (T) | |
| 1-Aza-12-crown 4-Ether |
| 58°C | |
| MFCD00134269 | |
| 1-aza-12-crown 4-ether, aza-12-crown-4, acmc-209spc, 1-aza-12-crown4-ether, 1-aza-4,7,10-trioxacyclododecane, 1,4,7-trioxa-10-aza cyclododecane, 1-aza-12-crown | |
| C1COCCOCCOCC[NH2+]1 | |
| 176.24 | |
| 175.23 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes