accessibility menu, dialog, popup

UserName

CAS RN 41775-76-2

OOOH2N

CAS RN 41775-76-2

IUPAC Name: 1,4,7-trioxa-10-azacyclododecan-10-ium
Synonyms: 1,4,7-trioxa-10-azacyclododecan-10-ium
Molecular Weight (g/mol): 176.24
Molecular Formula: C8H18NO3
InChi Key: QNSRHBOZQLXYNV-UHFFFAOYSA-O
SMILES: C1COCCOCCOCC[NH2+]1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results