missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Aminotridecane 98.0+%, TCI America™
$231.14 - $417.56
Chemical Identifiers
| CAS | 2869-34-3 |
|---|---|
| Molecular Formula | C13H29N |
| Molecular Weight (g/mol) | 199.382 |
| MDL Number | MFCD00008156 |
| InChI Key | ABVVEAHYODGCLZ-UHFFFAOYSA-N |
| Synonym | 1-aminotridecane, 1-tridecanamine, tridecylamine, n-tridecylamine, monotridecylamine, tridecanamine, 1-tridecylamine, unii-5s0kq9wz2d, 5s0kq9wz2d, tridecyl-amine |
| PubChem CID | 40761 |
| IUPAC Name | tridecan-1-amine |
| SMILES | CCCCCCCCCCCCCN |
Chemical Identifiers
| 2869-34-3 | |
| 199.382 | |
| ABVVEAHYODGCLZ-UHFFFAOYSA-N | |
| 40761 | |
| CCCCCCCCCCCCCN |
| C13H29N | |
| MFCD00008156 | |
| 1-aminotridecane, 1-tridecanamine, tridecylamine, n-tridecylamine, monotridecylamine, tridecanamine, 1-tridecylamine, unii-5s0kq9wz2d, 5s0kq9wz2d, tridecyl-amine | |
| tridecan-1-amine |
Specifications
| 2869-34-3 | |
| 275°C | |
| MFCD00008156 | |
| 3259 | |
| ABVVEAHYODGCLZ-UHFFFAOYSA-N | |
| tridecan-1-amine | |
| 40761 | |
| ≥98.0% (GC,T) | |
| 1-Aminotridecane |
| White-Yellow | |
| C13H29N | |
| 10 g | |
| 1-aminotridecane, 1-tridecanamine, tridecylamine, n-tridecylamine, monotridecylamine, tridecanamine, 1-tridecylamine, unii-5s0kq9wz2d, 5s0kq9wz2d, tridecyl-amine | |
| CCCCCCCCCCCCCN | |
| 199.382 | |
| 199.38 | |
| Crystals |
Safety and Handling
EINECSNumber : (2)-0133
RTECSNumber : YD3011650
TSCA : Yes