accessibility menu, dialog, popup

UserName

CAS RN 2869-34-3

H3CNH2

CAS RN 2869-34-3

IUPAC Name: tridecan-1-amine
Synonyms: 1-tridecanamine
Molecular Weight (g/mol): 199.38
Molecular Formula: C13H29N
InChi Key: ABVVEAHYODGCLZ-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCN
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results