missing translation for 'onlineSavingsMsg'
Learn More

1-Aminopyrrole, Thermo Scientific Chemicals

Catalog No. ac467560010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC467560010 1 g
AC467560050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC467560010 Supplier Thermo Scientific Chemicals Supplier No. 467560010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 765-39-9
Molecular Formula C4H6N2
Molecular Weight (g/mol) 82.11
InChI Key YNZAFFFENDLJQG-UHFFFAOYSA-N
IUPAC Name 1H-pyrrol-1-amine
SMILES NN1C=CC=C1

Specifications

Density 1.0600 g/mL
Boiling Point 174.0°C
Quantity 1 g
Specific Gravity 1.06
Formula Weight 82.1
Chemical Name or Material 1-Aminopyrrole
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Causes severe skin burns and eye damage.

Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection.

IF SWALLOWED: Rinse mouth.

Do NOT induce vomiting.

IF ON SKIN (or hair): Take off immediately all contaminated clothing.

Rinse skin with water or shower IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

Immediately call a POISON CENTER or doctor/physician.

RUO – Research Use Only