accessibility menu, dialog, popup

UserName

CAS RN 765-39-9

IUPAC Name: 1H-pyrrol-1-amine
Synonyms: pyrrol-1-amine
Molecular Weight (g/mol): 82.11
Molecular Formula: C4H6N2
InChi Key: YNZAFFFENDLJQG-UHFFFAOYSA-N
SMILES: NN1C=CC=C1
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)