missing translation for 'onlineSavingsMsg'
Learn More

1-Aminopyrrole 98.0+%, TCI America™

Catalog No. A10221G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A10221G 1 g
A10225G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A10221G Supplier TCI America Supplier No. A10221G

Chemical Identifiers

CAS 765-39-9
Molecular Formula C4H6N2
Molecular Weight (g/mol) 82.106
MDL Number MFCD00059931
InChI Key YNZAFFFENDLJQG-UHFFFAOYSA-N
PubChem CID 136589
IUPAC Name pyrrol-1-amine
SMILES C1=CN(C=C1)N

Specifications

Color Yellow
Boiling Point 174°C
Quantity 1 g
UN Number 3267
Formula Weight 82.11
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-Aminopyrrole
TSCA No
Recommended Storage Refrigerator