missing translation for 'onlineSavingsMsg'
Learn More

1-Aminopiperidine 98.0+%, TCI America™

Supplier:  TCI America A224125G

 View more versions of this product

Catalog No. A224125G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1-Aminopiperidine
Yellow
C5H12N2
25 g
1-aminopiperidine, n-aminopiperidine, 1-piperidinamine, piperidine, 1-amino, n-piperidylamine, aminopiperidine, piperidinamine, 1,1-pentamethylenehydrazine, piperidylamine, piperidine amine
C1CCN(CC1)N
100.165
100.17
Liquid
2213-43-6
146°C
MFCD00006489
1993
LWMPFIOTEAXAGV-UHFFFAOYSA-N
piperidin-1-amine
16658
≥98.0% (GC,T)

Chemical Identifiers

2213-43-6
100.165
LWMPFIOTEAXAGV-UHFFFAOYSA-N
16658
C1CCN(CC1)N
C5H12N2
MFCD00006489
1-aminopiperidine, n-aminopiperidine, 1-piperidinamine, piperidine, 1-amino, n-piperidylamine, aminopiperidine, piperidinamine, 1,1-pentamethylenehydrazine, piperidylamine, piperidine amine
piperidin-1-amine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : TM4165000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions