missing translation for 'onlineSavingsMsg'
Learn More

1-Aminopiperidine 98.0+%, TCI America™

Catalog No. p-7117798
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A22415G 5 g
A224125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A22415G Supplier TCI America Supplier No. A22415G
Only null left

Chemical Identifiers

CAS 2213-43-6
Molecular Formula C5H12N2
Molecular Weight (g/mol) 100.165
MDL Number MFCD00006489
InChI Key LWMPFIOTEAXAGV-UHFFFAOYSA-N
Synonym 1-aminopiperidine, n-aminopiperidine, 1-piperidinamine, piperidine, 1-amino, n-piperidylamine, aminopiperidine, piperidinamine, 1,1-pentamethylenehydrazine, piperidylamine, piperidine amine
PubChem CID 16658
IUPAC Name piperidin-1-amine
SMILES C1CCN(CC1)N

Specifications

Color Yellow
Boiling Point 146°C
Quantity 5 g
UN Number 1993
Formula Weight 100.17
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-Aminopiperidine
RTECSNumber TM4165000
TSCA No