missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Aminopentadecane 95.0+%, TCI America™
$404.51 - $747.49
Chemical Identifiers
| CAS | 2570-26-5 |
|---|---|
| Molecular Formula | C15H33N |
| Molecular Weight (g/mol) | 227.436 |
| MDL Number | MFCD00038195 |
| InChI Key | JPZYXGPCHFZBHO-UHFFFAOYSA-N |
| Synonym | pentadecylamine, 1-aminopentadecane, n-pentadecylamine, 1-pentadecanamine, 1-pentadecylamine, pentadecane, 1-amino, unii-2f7n79m9vh, acmc-209gly, 4-04-00-00817 beilstein handbook reference |
| PubChem CID | 17386 |
| ChEBI | CHEBI:87551 |
| IUPAC Name | pentadecan-1-amine |
| SMILES | CCCCCCCCCCCCCCCN |
Chemical Identifiers
| 2570-26-5 | |
| 227.436 | |
| JPZYXGPCHFZBHO-UHFFFAOYSA-N | |
| 17386 | |
| pentadecan-1-amine |
| C15H33N | |
| MFCD00038195 | |
| pentadecylamine, 1-aminopentadecane, n-pentadecylamine, 1-pentadecanamine, 1-pentadecylamine, pentadecane, 1-amino, unii-2f7n79m9vh, acmc-209gly, 4-04-00-00817 beilstein handbook reference | |
| CHEBI:87551 | |
| CCCCCCCCCCCCCCCN |
Specifications
| 2570-26-5 | |
| White | |
| MFCD00038195 | |
| 2923 | |
| JPZYXGPCHFZBHO-UHFFFAOYSA-N | |
| pentadecan-1-amine | |
| 17386 | |
| 227.44 | |
| Crystalline Powder |
| 37°C | |
| C15H33N | |
| 5 g | |
| pentadecylamine, 1-aminopentadecane, n-pentadecylamine, 1-pentadecanamine, 1-pentadecylamine, pentadecane, 1-amino, unii-2f7n79m9vh, acmc-209gly, 4-04-00-00817 beilstein handbook reference | |
| CCCCCCCCCCCCCCCN | |
| 227.436 | |
| CHEBI:87551 | |
| ≥95.0% (GC,T) | |
| 1-Aminopentadecane |
Safety and Handling
EINECSNumber : (2)-0133
RTECSNumber : RZ2120000
TSCA : Yes