accessibility menu, dialog, popup

UserName

CAS RN 2570-26-5

H3CNH2

CAS RN 2570-26-5

IUPAC Name: pentadecan-1-amine
Synonyms: pentadecylamine
Molecular Weight (g/mol): 227.44
Molecular Formula: C15H33N
InChi Key: JPZYXGPCHFZBHO-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCN
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results