missing translation for 'onlineSavingsMsg'
Learn More

1-Aminoindan 98.0+%, TCI America™

Catalog No. A130910G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A130910G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A130910G Supplier TCI America Supplier No. A130910G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 34698-41-4
Molecular Formula C9H12N
Molecular Weight (g/mol) 134.20
MDL Number MFCD00003799
InChI Key XJEVHMGJSYVQBQ-VIFPVBQESA-O
Synonym 1-aminoindan, 1-aminoindane, 1-indanamine, indan-1-ylamine, indanamin, aminoindan, indanylamine, indan-1-amine, 1h-inden-1-amine, 2,3-dihydro, 2,3-dihydro-1h-inden-1-ylamine
PubChem CID 123445
IUPAC Name (1S)-2,3-dihydro-1H-inden-1-aminium
SMILES [NH3+][C@H]1CCC2=CC=CC=C12

Specifications

Color Yellow
Boiling Point 220°C
Quantity 10 g
Formula Weight 133.19
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-Aminoindan