missing translation for 'onlineSavingsMsg'
Learn More

1-Aminoindan, 98%

Catalog No. p-56623
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC103850010 1 g Glass bottle
AC103850050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC103850010 Supplier Thermo Scientific Chemicals Supplier No. 103850010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 34698-41-4 | C9H12N | 134.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 34698-41-4
Molecular Formula C9H12N
Molecular Weight (g/mol) 134.20
MDL Number MFCD00003799
InChI Key XJEVHMGJSYVQBQ-VIFPVBQESA-O
Synonym 1-aminoindan, 1-aminoindane, 1-indanamine, indan-1-ylamine, indanamin, aminoindan, indanylamine, indan-1-amine, 1h-inden-1-amine, 2,3-dihydro, 2,3-dihydro-1h-inden-1-ylamine
PubChem CID 123445
IUPAC Name 2,3-dihydro-1H-inden-1-amine
SMILES [NH3+][C@H]1CCC2=CC=CC=C12

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 1.5°C
Color Colorless to Green
Density 1.0380g/mL
Boiling Point 96°C to 97°C (8.0 mmHg)
Flash Point 94°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5600 to 1.564
Quantity 1 g
Beilstein 12, 1191
Specific Gravity 1.038
Solubility Information Solubility in water: insoluble
Formula Weight 133.19
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Aminoindan, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 252-158-7
TSCA TSCA
Recommended Storage Product darkens on storage

RUO – Research Use Only