missing translation for 'onlineSavingsMsg'
Learn More

1-Aminohydantoin Hydrochloride 98.0+%, TCI America™

Supplier:  TCI America A190225G

 View more versions of this product

Catalog No. A190225G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1-Aminohydantoin Hydrochloride
203°C
C3H6ClN3O2
1-aminohydantoin hydrochloride, 1-aminoimidazolidine-2,4-dione hydrochloride, 1-aminohydantoin hcl, 1-amino hydantoin hydrochloride, 2,4-imidazolidinedione, 1-amino-, monohydrochloride, acmc-209h1m, 1-aminoimidazolidine-2,4-dione hcl, 1-aminoimidazoline-2,4-dione hydrochloride
C1C(=O)NC(=O)N1N.Cl
151.55
151.55
Crystalline Powder
2827-56-7
White-Yellow
25 g
WEOHANUVLKERQI-UHFFFAOYSA-N
1-aminoimidazolidine-2,4-dione;hydrochloride
12472963
≥98.0% (T)

Chemical Identifiers

2827-56-7
151.55
1-aminohydantoin hydrochloride, 1-aminoimidazolidine-2,4-dione hydrochloride, 1-aminohydantoin hcl, 1-amino hydantoin hydrochloride, 2,4-imidazolidinedione, 1-amino-, monohydrochloride, acmc-209h1m, 1-aminoimidazolidine-2,4-dione hcl, 1-aminoimidazoline-2,4-dione hydrochloride
1-aminoimidazolidine-2,4-dione;hydrochloride
C3H6ClN3O2
WEOHANUVLKERQI-UHFFFAOYSA-N
12472963
C1C(=O)NC(=O)N1N.Cl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions