missing translation for 'onlineSavingsMsg'
Learn More

1-Aminohydantoin Hydrochloride 98.0+%, TCI America™

Catalog No. p-7117625
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A19025G 5 g
A190225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A19025G Supplier TCI America Supplier No. A19025G
Only null left

Chemical Identifiers

CAS 2827-56-7
Molecular Formula C3H6ClN3O2
Molecular Weight (g/mol) 151.55
InChI Key WEOHANUVLKERQI-UHFFFAOYSA-N
Synonym 1-aminohydantoin hydrochloride, 1-aminoimidazolidine-2,4-dione hydrochloride, 1-aminohydantoin hcl, 1-amino hydantoin hydrochloride, 2,4-imidazolidinedione, 1-amino-, monohydrochloride, acmc-209h1m, 1-aminoimidazolidine-2,4-dione hcl, 1-aminoimidazoline-2,4-dione hydrochloride
PubChem CID 12472963
IUPAC Name 1-aminoimidazolidine-2,4-dione;hydrochloride
SMILES C1C(=O)NC(=O)N1N.Cl

Specifications

Melting Point 203°C
Color White-Yellow
Quantity 5 g
Formula Weight 151.55
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Aminohydantoin Hydrochloride