missing translation for 'onlineSavingsMsg'
Learn More

1-Aminocyclopropanecarbonitrile Hydrochloride 98.0+%, TCI America™

Catalog No. a24935g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
A24935G 5 g
A24931G 1 g
2 options

Catalog No. A24935G

Supplier: TCI America A24935G

Only null left

Chemical Identifiers

CAS 127946-77-4
Molecular Formula C4H7ClN2
Molecular Weight (g/mol) 118.564
MDL Number MFCD04114063
InChI Key PCEIEQLJYDMRFZ-UHFFFAOYSA-N
Synonym 1-amino-1-cyclopropanecarbonitrile hydrochloride, 1-aminocyclopropanecarbonitrile hydrochloride, 1-aminocyclopropane-1-carbonitrile hydrochloride, 1-aminocyclopropanecarbonitrile hcl, 1-amino-cyclopropyl cyanic hydrochloride, 1-amino-1-cyanocyclopropane hydrochloride, 1-amino-cyclopropanecarbonitrile hydrochloride, cyclopropanecarbonitrile, 1-amino-, monohydrochloride, 1-cyanocyclopropanaminium chloride, pubchem16832
PubChem CID 14586363
IUPAC Name 1-aminocyclopropane-1-carbonitrile;hydrochloride
SMILES C1CC1(C#N)N.Cl

Specifications

Melting Point 220°C
Color White-Yellow
Quantity 5 g
UN Number 3439
Formula Weight 118.56
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Aminocyclopropanecarbonitrile Hydrochloride