missing translation for 'onlineSavingsMsg'
Learn More

1-Aminocyclopentanecarboxylic Acid 98.0+%, TCI America™

Catalog No. p-7117220
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A10631G 1 g
A10635G 5 g
A106325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A10631G Supplier TCI America Supplier No. A10631G
Only null left

Chemical Identifiers

CAS 52-52-8
Molecular Formula C6H11NO2
Molecular Weight (g/mol) 129.159
MDL Number MFCD00001381
InChI Key NILQLFBWTXNUOE-UHFFFAOYSA-N
Synonym cycloleucine, 1-aminocyclopentanecarboxylic acid, cycloleucin, 1-amino-1-cyclopentanecarboxylic acid, 1-amino-1-carboxycyclopentane, 1-amino-cyclopentanecarboxylic acid, cyclopentanecarboxylic acid, 1-amino, cyclo-leucine, unii-0tqu7668ei, 1-aminocyclopentanecarboxylate
PubChem CID 2901
ChEBI CHEBI:40547
IUPAC Name 1-aminocyclopentane-1-carboxylic acid
SMILES C1CCC(C1)(C(=O)O)N

Specifications

Melting Point 330°C
Color White
Quantity 1 g
UN Number 2811
Formula Weight 129.16
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Aminocyclopentanecarboxylic Acid
RTECSNumber GY2625000
TSCA Yes