accessibility menu, dialog, popup

UserName

H2NHOO

CAS RN 52-52-8

IUPAC Name: 1-aminocyclopentane-1-carboxylic acid
Synonyms: cycloleucine acid, aminocyclopentanecarboxylic cyclo-leucine 1-aminocyclopentanecarboxylate ACPC cycloleucin
Molecular Weight (g/mol): 129.16
Molecular Formula: C6H11NO2
InChi Key: NILQLFBWTXNUOE-UHFFFAOYSA-N
SMILES: NC1(CCCC1)C(O)=O
Molecular Weight (g/mol) 
  • (5)
  • (1)
Percent Purity 
  • (2)
  • (3)
  • (1)
Quantity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (5)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)