missing translation for 'onlineSavingsMsg'
Learn More

1-Aminocyclopentanecarboxylic acid, 97+%

Catalog No. AAB2217903
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2217903 1 g
AAB2217914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2217903 Supplier Thermo Scientific Chemicals Supplier No. B2217903

CAS: 52-52-8 | C6H11NO2 | 129.159 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 52-52-8
Molecular Formula C6H11NO2
Molecular Weight (g/mol) 129.159
MDL Number MFCD00001381
InChI Key NILQLFBWTXNUOE-UHFFFAOYSA-N
Synonym cycloleucine, 1-aminocyclopentanecarboxylic acid, cycloleucin, 1-amino-1-cyclopentanecarboxylic acid, 1-amino-1-carboxycyclopentane, 1-amino-cyclopentanecarboxylic acid, cyclopentanecarboxylic acid, 1-amino, cyclo-leucine, unii-0tqu7668ei, 1-aminocyclopentanecarboxylate
PubChem CID 2901
ChEBI CHEBI:40547
IUPAC Name 1-aminocyclopentane-1-carboxylic acid
SMILES C1CCC(C1)(C(=O)O)N

Specifications

Melting Point ∼320°C (decomposition)
Quantity 1 g
UN Number UN2811
Beilstein 636626
Merck Index 14,2734
Formula Weight 129.16
Percent Purity ≥97%
Chemical Name or Material 1-Aminocyclopentanecarboxylic acid
Hazard Category H301
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 200-144-6
RTECSNumber GY2625000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only