missing translation for 'onlineSavingsMsg'
Learn More

1-Aminoanthracene 98.0+%, TCI America™

Catalog No. a17091g
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A17091G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A17091G Supplier TCI America Supplier No. A17091G
Only null left

Chemical Identifiers

CAS 610-49-1
Molecular Formula C14H11N
Molecular Weight (g/mol) 193.25
MDL Number MFCD00003579
InChI Key YUENFNPLGJCNRB-UHFFFAOYSA-N
Synonym 1-aminoanthracene, 1-anthramine, 1-anthracenamine, 1-anthrylamine, 1-anthracylamine, anthracenamine, alpha-aminoanthracene, anthracen-1-ylamine, anthracene, amino, aminoanthracene
PubChem CID 11885
ChEBI CHEBI:40678
IUPAC Name anthracen-1-amine
SMILES NC1=C2C=C3C=CC=CC3=CC2=CC=C1

Specifications

Melting Point 121°C
Quantity 1 g
Formula Weight 193.25
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Aminoanthracene
RTECSNumber CA9100000
TSCA No