CAS RN 610-49-1
CAS RN 610-49-1
IUPAC Name:
anthracen-1-amine
Synonyms:
aminoanthracene amino-anthracene 1-anthracylamine 1-anthramine (6CI,7CI,8CI) ccris 749 anthramine anthracene, amino-
Molecular Weight (g/mol):
193.25
Molecular Formula:
C14H11N
InChi Key:
YUENFNPLGJCNRB-UHFFFAOYSA-N
SMILES:
NC1=C2C=C3C=CC=CC3=CC2=CC=C1
1
–
1
of
1
results
1
1-Aminoanthracene 98.0+%, TCI America™
CAS: 610-49-1 Molecular Formula: C14H11N Molecular Weight (g/mol): 193.25 MDL Number: MFCD00003579 InChI Key: YUENFNPLGJCNRB-UHFFFAOYSA-N Synonym: 1-aminoanthracene,1-anthramine,1-anthracenamine,1-anthrylamine,1-anthracylamine,anthracenamine,alpha-aminoanthracene,anthracen-1-ylamine,anthracene, amino,aminoanthracene PubChem CID: 11885 ChEBI: CHEBI:40678 IUPAC Name: anthracen-1-amine SMILES: NC1=C2C=C3C=CC=CC3=CC2=CC=C1