missing translation for 'onlineSavingsMsg'
Learn More

1-Amino-4-oxocyclohexanecarboxylic acid ethylene ketal, 98%, Thermo Scientific Chemicals

Catalog No. p-7047813
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1979503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1979503 Supplier Thermo Scientific Chemicals Supplier No. L1979503

1-Amino-4-oxocyclohexanecarboxylic acid ethylene ketal is used in modification of polymer properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Amino-4-oxocyclohexanecarboxylic acid ethylene ketal is used in modification of polymer properties.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 54621-18-0
Molecular Formula C9H15NO4
Molecular Weight (g/mol) 201.222
MDL Number MFCD03790906
InChI Key HIIFALCQUKTUHB-UHFFFAOYSA-N
Synonym 8-amino-1,4-dioxaspiro 4.5 decane-8-carboxylic acid, 1-amino-4-oxocyclohexanecarboxylic acid ethylene ketal, 1-amino-4-oxocyclohexanecarboxylicacidethyleneketal, 1-amino-4,4-ethylenedioxycyclohexane carboxylic acid, 4-amino-4-carboxycyclohexanone ethylene ketal, 1,4-dioxaspiro 4.5 decane-8-carboxylicacid, 8-amino, 4-amino-4-2-hydroxy-1,3-dioxolan-2-yl cyclohexanone, 8-amino-1,4-dioxa-spiro 4.5 decane-8-carboxylic acid, amino-4-ethylene-ketal cyclohexylcarboxylic acid, 1-amino-4-oxocyclohexanecarboxylic acid ethylene k
PubChem CID 7015505
IUPAC Name 8-amino-1,4-dioxaspiro[4.5]decane-8-carboxylic acid
SMILES C1CC2(CCC1(C(=O)O)N)OCCO2

Specifications

Melting Point 301°C to 304°C
Quantity 1 g
Beilstein 1373325
Solubility Information Insoluble in water.
Formula Weight 201.22
Percent Purity 98%
Chemical Name or Material 1-Amino-4-oxocyclohexanecarboxylic acid ethylene ketal
EINECSNumber 000-000-0
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only