accessibility menu, dialog, popup

UserName

OOHNH2OO

CAS RN 54621-18-0

IUPAC Name: 8-amino-1,4-dioxaspiro[4.5]decane-8-carboxylic acid
Synonyms: 8-amino-1,4-dioxaspiro[4.5]decane-8-carboxylic acid
Molecular Weight (g/mol): 201.22
Molecular Formula: C9H15NO4
InChi Key: HIIFALCQUKTUHB-UHFFFAOYSA-N
SMILES: NC1(CCC2(CC1)OCCO2)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results