missing translation for 'onlineSavingsMsg'
Learn More

1-Amino-2-napthol-4-sulfonic Acid, ACS Reagent Grade, Ricca Chemical

Catalog No. RDCA035025F
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RDCA035025F 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RDCA035025F Supplier Ricca Chemical Company Supplier No. RDCA035025F1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 116-63-2
Molecular Formula C10H9NO4S
Molecular Weight (g/mol) 239.245
InChI Key RXCMFQDTWCCLBL-UHFFFAOYSA-N
Synonym 1-amino-2-naphthol-4-sulfonic acid, 4-amino-3-hydroxy-1-naphthalenesulfonic acid, 1-amino-4-sulfo-2-naphthol, 1,2,4-acid, unii-w46sk3p33g, 1-naphthalenesulfonic acid, 4-amino-3-hydroxy, 4-amino-3-hydroxynaphthalene-1-sulphonic acid, 4-amino-3-hydroxynaphthalenesulfonic acid, 1,2,4-aminonaphtholsulfonic acid, 2-hydroxy-4-sulfo-1-naphthylamine
PubChem CID 8316
ChEBI CHEBI:19024
IUPAC Name 4-amino-3-hydroxynaphthalene-1-sulfonic acid
SMILES C1=CC=C2C(=C1)C(=CC(=C2N)O)S(=O)(=O)O

Specifications

Color Pink to White
CAS Min % 98.0
CAS Max % 100.0
Percent Purity Min. 90.%
Grade ACS
Concentration or Composition (by Analyte or Components) 1-Amino-2-Napthol-4-sulfonic Acid
Packaging Pouch
Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material 1-Amino-2-Napthol-4-sulfonic Acid
ShelfLife 60 months