missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Amino-2-Naphthol-4-Sulfonic Acid Solution, For Silica, Certified, LabChem™
Supplier: LabChem LC108901
Specifications
| 1-Amino-2-Naphthol-4-Sulfonic Acid Solution | |
| For Silica | |
| 84.25,15,0.5,0.25 | |
| water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide | |
| 239.26 | |
| Passes Test | |
| Passes Test | |
| 500 mL |
| Brown | |
| 7732-18-5,7631-90-5,7757-83-7,116-63-2 | |
| H2N(HO)C10H5SO3H | |
| Soluble in water | |
| Certified | |
| Amber Poly | |
| Sulfur compounds; Carbon monoxide; Carbon dioxide | |
| Liquid |
Chemical Identifiers
| 7732-18-5 | |
| water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/at
Warning
Recommended Storage : Room Temperature