missing translation for 'onlineSavingsMsg'
Learn More

1-Amino-2-methylpyridinium Iodide 98.0+%, TCI America™

Catalog No. A25581G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A25581G 1 g
A25585G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A25581G Supplier TCI America Supplier No. A25581G
Only null left

Chemical Identifiers

CAS 7583-90-6
Molecular Formula C6H9IN2
Molecular Weight (g/mol) 236.056
MDL Number MFCD00955862
InChI Key XMFXSCPQGJZSLU-UHFFFAOYSA-M
PubChem CID 12291507
IUPAC Name 2-methylpyridin-1-ium-1-amine;iodide
SMILES CC1=CC=CC=[N+]1N.[I-]

Specifications

Melting Point 152°C
Color White-Yellow
Quantity 1 g
Formula Weight 236.06
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Amino-2-methylpyridinium Iodide