accessibility menu, dialog, popup

UserName

CH3NH2NI

CAS RN 7583-90-6

IUPAC Name: 1-amino-2-methylpyridin-1-ium iodide
Synonyms: 1-amino-2-methylpyridin-1-ium iodide
Molecular Weight (g/mol): 236.06
Molecular Formula: C6H9IN2
InChi Key: XMFXSCPQGJZSLU-UHFFFAOYSA-M
SMILES: [I-].CC1=CC=CC=[N+]1N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results