missing translation for 'onlineSavingsMsg'
Learn More

1-Amino-2-methylnaphthalene 98.0+%, TCI America™

Catalog No. p-7117128
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A09175G 5 g
A091725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A09175G Supplier TCI America Supplier No. A09175G

Chemical Identifiers

CAS 2246-44-8
Molecular Formula C11H11N
Molecular Weight (g/mol) 157.216
MDL Number MFCD00059135
InChI Key JMBLSGAXSMOKPN-UHFFFAOYSA-N
Synonym 2-Methyl-1-naphthylamine
PubChem CID 16733
IUPAC Name 2-methylnaphthalen-1-amine
SMILES CC1=C(C2=CC=CC=C2C=C1)N

Specifications

Quantity 5 g
Formula Weight 157.22
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 1-Amino-2-methylnaphthalene
RTECSNumber QM3675000
TSCA No
Recommended Storage Refrigerator